missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-8-quinolinol, TCI America™

Manufacturer:  TCI America M0420500G

 View more versions of this product

Catalog No. M0420500G



Specifications

Specifications

2-Methyl-8-quinolinol
Undesignated
500g
C10H9NO
3077
NBYLBWHHTUWMER-UHFFFAOYSA-N
2-methylquinolin-8-ol
13224
Crystal-Powder at 20°C
145°C
73°C
826-81-3
MFCD00006765
8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol
CC1=NC2=C(C=CC=C2O)C=C1
159.188
159.19
≥98.0% (GC,T)

Chemical Identifiers

826-81-3
159.188
NBYLBWHHTUWMER-UHFFFAOYSA-N
13224
CC1=NC2=C(C=CC=C2O)C=C1
C10H9NO
MFCD00006765
8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol
2-methylquinolin-8-ol
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0808

RTECSNumber : VC7920000

TSCA : Yes

Documents