missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-7-[phenyl(phenylamino)methyl]-8-quinolinol 98.0+%, TCI America™

$431.22 - $1,285.42

Chemical Identifiers

CAS 6964-62-1
Molecular Formula C23H20N2O
Molecular Weight (g/mol) 340.426
MDL Number MFCD08823776
InChI Key WEENRMPCSWFMTE-UHFFFAOYSA-N
Synonym 2-methyl-7-phenyl phenylamino methyl quinolin-8-ol, 2-methyl-7-phenyl phenylamino methyl-8-quinolinol, 2-methyl-7-phenyl-phenylamino methyl quinolin-8-ol, 7-anilino phenyl methyl-2-methyl-8-quinolinol, 7-anilino phenyl methyl-2-methylquinolin-8-ol, 7-anilino phenyl-methyl-2-methyl-8-quinolinol, 8-hydroxy-2-methyl-7-phenyl phenylamino methyl quinoline, 8-quinolinol,2-methyl-7-phenyl phenylamino methyl, d0c0tn
PubChem CID 248986
IUPAC Name 7-[anilino(phenyl)methyl]-2-methylquinolin-8-ol
SMILES CC1=NC2=C(C=C1)C=CC(=C2O)C(C3=CC=CC=C3)NC4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M239025MG View Documents Tci America
M239025MG
25mg Each for $431.22
 
M2390100MG View Documents Tci America
M2390100MG
100mg Each for $1,285.42
 
Specifications

Chemical Identifiers

6964-62-1
340.426
WEENRMPCSWFMTE-UHFFFAOYSA-N
248986
CC1=NC2=C(C=C1)C=CC(=C2O)C(C3=CC=CC=C3)NC4=CC=CC=C4
C23H20N2O
MFCD08823776
2-methyl-7-phenyl phenylamino methyl quinolin-8-ol, 2-methyl-7-phenyl phenylamino methyl-8-quinolinol, 2-methyl-7-phenyl-phenylamino methyl quinolin-8-ol, 7-anilino phenyl methyl-2-methyl-8-quinolinol, 7-anilino phenyl methyl-2-methylquinolin-8-ol, 7-anilino phenyl-methyl-2-methyl-8-quinolinol, 8-hydroxy-2-methyl-7-phenyl phenylamino methyl quinoline, 8-quinolinol,2-methyl-7-phenyl phenylamino methyl, d0c0tn
7-[anilino(phenyl)methyl]-2-methylquinolin-8-ol

Specifications

Undesignated
25mg
C23H20N2O
2-methyl-7-phenyl phenylamino methyl quinolin-8-ol, 2-methyl-7-phenyl phenylamino methyl-8-quinolinol, 2-methyl-7-phenyl-phenylamino methyl quinolin-8-ol, 7-anilino phenyl methyl-2-methyl-8-quinolinol, 7-anilino phenyl methyl-2-methylquinolin-8-ol, 7-anilino phenyl-methyl-2-methyl-8-quinolinol, 8-hydroxy-2-methyl-7-phenyl phenylamino methyl quinoline, 8-quinolinol,2-methyl-7-phenyl phenylamino methyl, d0c0tn
CC1=NC2=C(C=C1)C=CC(=C2O)C(C3=CC=CC=C3)NC4=CC=CC=C4
340.426
340.43
≥98.0% (GC)
145°C
6964-62-1
MFCD08823776
WEENRMPCSWFMTE-UHFFFAOYSA-N
7-[anilino(phenyl)methyl]-2-methylquinolin-8-ol
248986
Crystal-Powder at 20°C
2-Methyl-7-[phenyl(phenylamino)methyl]-8-quinolinol
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator