missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-6-nitroquinoline, 97%, Maybridge

Manufacturer:  Thermo Scientific BTB06234EA

 View more versions of this product

Catalog No. BTB06234EA



Specifications

Specifications

2-Methyl-6-nitroquinoline
10g
96.5
97%
MFCD00051736
DXDPHHQJZWWAEH-UHFFFAOYSA-N
2-methyl-6-nitroquinoline
69172
97%
Amber Glass Bottle
613-30-9
100.0
C10H8N2O2
1-2-aminoethyl pyrrole-2,5-dione,hydrochloride
CC1=NC2=C(C=C1)C=C(C=C2)[N+](=O)[O-]
188.186
188.19

Chemical Identifiers

613-30-9
188.186
DXDPHHQJZWWAEH-UHFFFAOYSA-N
69172
CC1=NC2=C(C=C1)C=C(C=C2)[N+](=O)[O-]
C10H8N2O2
MFCD00051736
1-2-aminoethyl pyrrole-2,5-dione,hydrochloride
2-methyl-6-nitroquinoline
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents