Please login to your online account to display your discounted pricing

2-Methyl-6-nitroquinoline, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB06234DA

 View more versions of this product

Catalog No. BTB06234DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 188.19
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2-Methyl-6-nitroquinoline
MDL Number MFCD00051736
CAS 613-30-9
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H8N2O2
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Methyl-6-nitroquinoline
CAS 613-30-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2O2
Formula Weight 188.19
MDL Number MFCD00051736
Accessories

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.