missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-6-nitrobenzoyl Chloride, TCI America™

$151.25 - $389.59

Chemical Identifiers

CAS 66232-57-3
Molecular Formula C8H6ClNO3
Molecular Weight (g/mol) 199.59
MDL Number MFCD06797116
InChI Key ZMAJSTDJAIVHGZ-UHFFFAOYSA-N
Synonym 6-Nitro-o-toluoyl Chloride
PubChem CID 12819896
IUPAC Name 2-methyl-6-nitrobenzoyl chloride
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14381G View Documents TCI America
M14381G
1g Each for $151.25
 
M14385G View Documents TCI America
M14385G
5g Each for $389.59
 
Specifications

Chemical Identifiers

66232-57-3
199.59
ZMAJSTDJAIVHGZ-UHFFFAOYSA-N
12819896
CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)Cl
C8H6ClNO3
MFCD06797116
6-Nitro-o-toluoyl Chloride
2-methyl-6-nitrobenzoyl chloride

Specifications

Undesignated
1g
C8H6ClNO3
3261
ZMAJSTDJAIVHGZ-UHFFFAOYSA-N
2-methyl-6-nitrobenzoyl chloride
12819896
Crystal-Powder at 20°C
2-Methyl-6-nitrobenzoyl Chloride
42°C
66232-57-3
MFCD06797116
6-Nitro-o-toluoyl Chloride
CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)Cl
199.59
199.59
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator