missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-6-nitrobenzoic acid, 99%, Acros Organics™

Manufacturer:  Thermo Fisher Scientific AC165980250

 View more versions of this product

Catalog No. AC165980250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methyl-6-nitrobenzoic acid
154.0°C to 157.0°C
13506-76-8
C8H7NO4
09,I,189
Solubility in water: <0.1g/100mL (20°C).
CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O
16097
99%
Authentic
25g
97.5% min. (HPLC)
CH3C6H3(NO2)CO2H
2-methyl-6-nitro-benzoic acid
CCXSGQZMYLXTOI-UHFFFAOYSA-N
181.15
181.15

Chemical Identifiers

13506-76-8
181.15
2-methyl-6-nitro-benzoic acid
CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O
C8H7NO4
CCXSGQZMYLXTOI-UHFFFAOYSA-N
16097
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 236-833-3

SDS Documents