Please login to your online account to display your discounted pricing

2-Methyl-6-nitroaniline 99%, ACROS Organics™

$172.54 - $172.54

Chemical Identifiers

Chemical Name or Material 2-Methyl-6-nitroaniline
Name Note 99%
CAS 570-24-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O2
Linear Formula CH3C6H3(NO2)NH2
Formula Weight 152.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC127230250

  SDS
thermo fisher scientific
127230250
Glass bottle 25g Each for $172.54

AC127230050

  SDS
thermo fisher scientific
AC127230050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC127231000

  SDS
thermo fisher scientific
AC127231000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 152.15
Color Brownish
Quantity 25g
Beilstein 12, 843
EINECS Number 209-329-6
TSCA TSCA
Melting Point 93° to 96°C
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 2-Amino-3-nitrotoluene
CAS Min % 98.5
Chemical Name or Material 2-Methyl-6-nitroaniline
Packaging Glass bottle
Name Note 99%
Molecular Formula C7H8N2O2
Linear Formula CH3C6H3(NO2)NH2
MDL Number MFCD00007744
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
CAS 570-24-1
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.