Please login to your online account to display your discounted pricing

2-Methyl-6-nitroaniline 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC127231000

 View more versions of this product

Catalog No. AC127231000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Brownish
Physical Form Crystalline Powder
Formula Weight 152.15
Assay ≥98.5%
Quantity 100g
TSCA TSCA
EINECS Number 209-329-6
Melting Point 93° to 96°C
Beilstein 12, 843
CAS Max % 100.0
Synonym 2-Amino-3-nitrotoluene
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 570-24-1
Assay Percent Range 99%
Name Note 99%
Chemical Name or Material 2-Methyl-6-nitroaniline
Linear Formula CH3C6H3(NO2)NH2
Molecular Formula C7H8N2O2
MDL Number MFCD00007744

Specifications

Chemical Name or Material 2-Methyl-6-nitroaniline
Name Note 99%
CAS 570-24-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O2
Linear Formula CH3C6H3(NO2)NH2
Formula Weight 152.15
MDL Number MFCD00007744
Synonym 2-Amino-3-nitrotoluene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents