Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Methyl-5'-(trifluoromethyl)acetophenone, 97%

Chemical Identifiers

Chemical Name or Material 2'-Methyl-5'-(trifluoromethyl)acetophenone
Molecular Formula C10H9F3O
Formula Weight 202.17
MDL Number MFCD09832290
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3181903

 
alfa aesar™
H3181903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3181906

 
alfa aesar™
H3181906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.17
Chemical Name or Material 2'-Methyl-5'-(trifluoromethyl)acetophenone
Assay 97%
Molecular Formula C10H9F3O
MDL Number MFCD09832290
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.