missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-propionyl-furan, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 233000500

 View more versions of this product

Catalog No. AC233000500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Methyl-5-propionyl-furan
95%
94.0
Authentic
1.5020 to 1.5050 (20°C, 589nm)
≥94%
C8H10O2
MFCD00053142
50g
10599-69-6
100.0
Glass bottle
Liquid
95%
138.17
Clear yellow

Chemical Identifiers

10599-69-6
100.0
C8H10O2
MFCD00053142
≥94%
94.0
95%
138.17
2-Methyl-5-propionyl-furan, 95%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 234-215-8

TSCA : TSCA