missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-propionyl-furan, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 233000100

 View more versions of this product

Catalog No. AC233000100


 

Specifications Description & Specifications

Specifications

≥94%
138.17
Clear yellow
100.0
Authentic
95%
2-Methyl-5-propionyl-furan
95%
Liquid
10g
94.0
1.5020 to 1.5050 (20°C, 589nm)
Glass bottle
10599-69-6
C8H10O2
MFCD00053142

Chemical Identifiers

10599-69-6
100.0
C8H10O2
MFCD00053142
≥94%
94.0
95%
138.17
2-Methyl-5-propionyl-furan, 95%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 234-215-8

TSCA : TSCA