Please login to your online account to display your discounted pricing

2-Methyl-5-propionyl-furan, 95%, ACROS Organics™

$165.19 - $165.19

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-propionyl-furan
Name Note 95%
CAS 10599-69-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H10O2
Formula Weight 138.17
MDL Number MFCD00053142
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC233000100

  SDS
thermo fisher scientific
233000100
Glass bottle 10g Each for $165.19

AC233000500

  SDS
thermo fisher scientific
233000500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 138.17
Quantity 10g
Color Clear yellow
TSCA TSCA
EINECS Number 234-215-8
CAS Min % 94.0
CAS Max % 100.0
Refractive Index 1.5020 to 1.5050 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%
CAS 10599-69-6
Chemical Name or Material 2-Methyl-5-propionyl-furan
Molecular Formula C8H10O2
Assay Percent Range 95%
MDL Number MFCD00053142