Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-phenylthiophene, 96%

$71.27 - $235.05

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-phenylthiophene
CAS 5069-26-1
Molecular Formula C11H10S
Formula Weight 174.27
MDL Number MFCD06201035
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2042003

  SDS
alfa aesar™
L2042003
1g Each for $71.27

AAL2042006

  SDS
alfa aesar™
L2042006
5g Each for $235.05
Description & Specifications

Specifications

Formula Weight 174.27
Chemical Name or Material 2-Methyl-5-phenylthiophene
CAS 5069-26-1
MDL Number MFCD06201035
Molecular Formula C11H10S
Odor Odorless
Quantity 1g
Melting Point 46° to 49°C
Assay 96%

Applications
2-Methyl-5-phenylthiophen is employed as intermediate for pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.