Please login to your online account to display your discounted pricing

2-Methyl-5-phenyl-3-furyl isocyanate, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC14906DA

 View more versions of this product

Catalog No. CC14906DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 199.21
Quantity 1g
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H9NO2
Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate
CAS 568577-82-2
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H9NO2
Formula Weight 199.21