Please login to your online account to display your discounted pricing

2-Methyl-5-phenyl-3-furyl isocyanate, Maybridge

$144.14 - $336.80

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H9NO2
Formula Weight 199.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC14906CB

  SDS
fisher scientific company
CC14906CB
Amber Glass Bottle 250mg Each for $144.14

CC14906DA

  SDS
fisher scientific uk ltd
CC14906DA
Amber Glass Bottle 1g Each for $336.80

CC14906DE

  SDS
fisher scientific company
CC14906DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.21
Quantity 250mg
Assay 95%
Assay Percent Range 95%
CAS 568577-82-2
Molecular Formula C12H9NO2
Packaging Amber Glass Bottle
Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate