Please login to your online account to display your discounted pricing

2-Methyl-5-phenyl-3-furyl isocyanate, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC14906DE

 View more versions of this product

Catalog No. CC14906DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 199.21
Quantity 5g
Assay 95%
CAS 568577-82-2
Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C12H9NO2

Specifications

Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H9NO2
Formula Weight 199.21