missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-phenyl-3-furoic acid, 97%, Maybridge

Manufacturer:  Thermo Scientific CC14901EA

 View more versions of this product

Catalog No. CC14901EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methyl-5-phenyl-3-furoic acid
10g
96.5
97%
MFCD00221070
VLMNACSEESRUAK-UHFFFAOYSA-N
2-methyl-5-phenylfuran-3-carboxylic acid
736516
97%
Amber Glass Bottle
108124-17-0
100.0
C12H10O3
2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid
CC1=C(C=C(O1)C2=CC=CC=C2)C(=O)O
202.209
202.21

Chemical Identifiers

108124-17-0
202.209
VLMNACSEESRUAK-UHFFFAOYSA-N
736516
CC1=C(C=C(O1)C2=CC=CC=C2)C(=O)O
C12H10O3
MFCD00221070
2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid
2-methyl-5-phenylfuran-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents