missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-oxo-1-cyclopentenyl Propionate, TCI America™

$235.28 - $817.38

Chemical Identifiers

CAS 87-55-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00798053
InChI Key NKRMCRMCMGRKHS-UHFFFAOYSA-N
Synonym Propionic Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
PubChem CID 66600
IUPAC Name (2-methyl-5-oxocyclopenten-1-yl) propanoate
SMILES CCC(=O)OC1=C(CCC1=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24875G View Documents TCI America
M24875G
5g Each for $235.28
 
M248725G View Documents TCI America
M248725G
25g Each for $817.38
 
Specifications

Chemical Identifiers

87-55-8
168.192
NKRMCRMCMGRKHS-UHFFFAOYSA-N
66600
CCC(=O)OC1=C(CCC1=O)C
C9H12O3
MFCD00798053
Propionic Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
(2-methyl-5-oxocyclopenten-1-yl) propanoate

Specifications

White-Yellow
87-55-8
MFCD00798053
NKRMCRMCMGRKHS-UHFFFAOYSA-N
(2-methyl-5-oxocyclopenten-1-yl) propanoate
66600
Crystal-Lump at 20°C
2-Methyl-5-oxo-1-cyclopentenyl Propionate
5g
C9H12O3
Propionic Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
CCC(=O)OC1=C(CCC1=O)C
168.192
168.19
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2383

TSCA : No