missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-5-oxo-1-cyclopentenyl Propionate 98.0+%, TCI America™
Supplier: TCI America M248725G
Specifications
2-Methyl-5-oxo-1-cyclopentenyl Propionate | |
White-Yellow | |
MFCD00798053 | |
Propionic Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester | |
CCC(=O)OC1=C(C)CCC1=O | |
168.19 | |
168.19 | |
Crystalline Lumps |
87-55-8 | |
C9H12O3 | |
25 g | |
NKRMCRMCMGRKHS-UHFFFAOYSA-N | |
2-methyl-5-oxocyclopent-1-en-1-yl propanoate | |
66600 | |
≥98.0% (GC) |
Chemical Identifiers
87-55-8 | |
168.19 | |
NKRMCRMCMGRKHS-UHFFFAOYSA-N | |
66600 | |
CCC(=O)OC1=C(C)CCC1=O |
C9H12O3 | |
MFCD00798053 | |
Propionic Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester | |
2-methyl-5-oxocyclopent-1-en-1-yl propanoate |
Safety and Handling
EINECSNumber : (3)-2383
TSCA : No