missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-oxo-1-cyclopentenyl Butyrate, TCI America™

$235.28 - $817.38

Chemical Identifiers

CAS 68227-51-0
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00798052
InChI Key SUJWBOKDKMEAOQ-UHFFFAOYSA-N
Synonym Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
PubChem CID 109828
IUPAC Name (2-methyl-5-oxocyclopenten-1-yl) butanoate
SMILES CCCC(=O)OC1=C(CCC1=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M248625G View Documents TCI America
M248625G
25g Each for $817.38
 
M24865G View Documents TCI America
M24865G
5g Each for $235.28
 
Specifications

Chemical Identifiers

68227-51-0
182.219
SUJWBOKDKMEAOQ-UHFFFAOYSA-N
109828
CCCC(=O)OC1=C(CCC1=O)C
C10H14O3
MFCD00798052
Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
(2-methyl-5-oxocyclopenten-1-yl) butanoate

Specifications

85°C
25g
C10H14O3
Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
CCCC(=O)OC1=C(CCC1=O)C
182.219
182.22
≥98.0% (GC)
Yellow
68227-51-0
MFCD00798052
SUJWBOKDKMEAOQ-UHFFFAOYSA-N
(2-methyl-5-oxocyclopenten-1-yl) butanoate
109828
Clear Liquid at 20°C
2-Methyl-5-oxo-1-cyclopentenyl Butyrate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2388

TSCA : Yes