Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-octyn-4-ol, 97%

$84.97 - $332.49

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-octyn-4-ol
CAS 60657-70-7
Molecular Formula C9H16O
Formula Weight 140.23
MDL Number MFCD00041690
Beilstein 8899472
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1262506

  SDS
alfa aesar™
L1262506
5g Each for $84.97

AAL1262514

  SDS
alfa aesar™
L1262514
25g Each for $332.49
Description & Specifications

Specifications

Formula Weight 140.23
Chemical Name or Material 2-Methyl-5-octyn-4-ol
Assay 97%
Molecular Formula C9H16O
MDL Number MFCD00041690
CAS 60657-70-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 8899472
Flash Point 78°C (172°F)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.