missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-octyn-4-ol, 97%

Manufacturer:  Alfa Aesar™ L1262514

 View more versions of this product

Catalog No. AAL1262514



Specifications

Specifications

2-Methyl-5-octyn-4-ol
25g
C9H16O
8899472
YBGQRUFRRPFNKW-UHFFFAOYSA-N
2-methyloct-5-yn-4-ol
143720
97%
78°C (172°F)
60657-70-7
MFCD00041690
2-methyl-5-octyn-4-ol, 5-octyn-4-ol, 2-methyl, acmc-20anql, 2-methyl-5-octyne-4-ol
CCC#CC(CC(C)C)O
140.226
140.23

Chemical Identifiers

60657-70-7
140.226
YBGQRUFRRPFNKW-UHFFFAOYSA-N
143720
CCC#CC(CC(C)C)O
C9H16O
MFCD00041690
2-methyl-5-octyn-4-ol, 5-octyn-4-ol, 2-methyl, acmc-20anql, 2-methyl-5-octyne-4-ol
2-methyloct-5-yn-4-ol
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents