missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-5-nitropyridine 96.0+%, TCI America™
$53.10 - $212.35
Chemical Identifiers
CAS | 21203-68-9 |
---|---|
Molecular Formula | C6H6N2O2 |
Molecular Weight (g/mol) | 138.126 |
MDL Number | MFCD04114179 |
InChI Key | USZINSZJSVMICC-UHFFFAOYSA-N |
Synonym | 5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036 |
PubChem CID | 2794552 |
IUPAC Name | 2-methyl-5-nitropyridine |
SMILES | CC1=NC=C(C=C1)[N+](=O)[O-] |
Chemical Identifiers
21203-68-9 | |
138.126 | |
USZINSZJSVMICC-UHFFFAOYSA-N | |
2794552 | |
CC1=NC=C(C=C1)[N+](=O)[O-] |
C6H6N2O2 | |
MFCD04114179 | |
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036 | |
2-methyl-5-nitropyridine |
Specifications
21203-68-9 | |
C6H6N2O2 | |
1 g | |
USZINSZJSVMICC-UHFFFAOYSA-N | |
2-methyl-5-nitropyridine | |
2794552 | |
≥96.0% (GC,T) | |
2-Methyl-5-nitropyridine |
109°C | |
MFCD04114179 | |
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036 | |
CC1=NC=C(C=C1)[N+](=O)[O-] | |
138.126 | |
138.13 | |
Crystalline Powder |
Safety and Handling
TSCA : No