Learn More
2-Methyl-5-nitropyridine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6175014
Description
2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2-Methyl-5-nitropyridine | |
C6H6N2O2 | |
25 g | |
Slightly soluble in water. | |
CC1=NC=C(C=C1)[N+](=O)[O-] | |
138.126 | |
138.12 | |
Solid |
21203-68-9 | |
MFCD04114179 | |
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036 | |
USZINSZJSVMICC-UHFFFAOYSA-N | |
2-methyl-5-nitropyridine | |
2794552 | |
95% |
Chemical Identifiers
21203-68-9 | |
138.126 | |
USZINSZJSVMICC-UHFFFAOYSA-N | |
2794552 | |
CC1=NC=C(C=C1)[N+](=O)[O-] |
C6H6N2O2 | |
MFCD04114179 | |
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036 | |
2-methyl-5-nitropyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only