Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitrophenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-nitrophenyl isocyanate
CAS 13471-68-6
Molecular Formula C8H6N2O3
Formula Weight 178.15
MDL Number MFCD00014718
UN Number UN2811
Beilstein 3272007
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1085003

  SDS
alfa aesar™
L1085003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1085006

  SDS
alfa aesar™
L1085006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.15
Chemical Name or Material 2-Methyl-5-nitrophenyl isocyanate
Assay 97%
Molecular Formula C8H6N2O3
MDL Number MFCD00014718
CAS 13471-68-6
UN Number UN2811
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H331-H334-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 3272007
Melting Point 49° to 52°C
Flash Point >110°C (230°F)
Odor Irritating
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C