missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitrophenyl isocyanate, 97%

Manufacturer:  Alfa Aesar™ L1085006

 View more versions of this product

Catalog No. AAL1085006



Specifications

Specifications

2-Methyl-5-nitrophenyl isocyanate
Irritating
5g
C8H6N2O3
UN2811
2-methyl-5-nitrophenyl isocyanate, benzene, 2-isocyanato-1-methyl-4-nitro, 2-methyl-5-nitrophenylisocyanate, acmc-1c1fb, 2-methyl-5-nitrobenzenisocyanate, benzene,2-isocyanato-1-methyl-4-nitro, 2-methyl-5-nitrophenyl isocyanate
CC1=C(C=C(C=C1)[N+](=O)[O-])N=C=O
178.147
178.15
>110°C (230°F)
49°C to 52°C
13471-68-6
MFCD00014718
3272007
PQFOUIMVTGBFRN-UHFFFAOYSA-N
2-isocyanato-1-methyl-4-nitrobenzene
292609
97%

Chemical Identifiers

13471-68-6
178.147
PQFOUIMVTGBFRN-UHFFFAOYSA-N
292609
CC1=C(C=C(C=C1)[N+](=O)[O-])N=C=O
C8H6N2O3
MFCD00014718
2-methyl-5-nitrophenyl isocyanate, benzene, 2-isocyanato-1-methyl-4-nitro, 2-methyl-5-nitrophenylisocyanate, acmc-1c1fb, 2-methyl-5-nitrobenzenisocyanate, benzene,2-isocyanato-1-methyl-4-nitro, 2-methyl-5-nitrophenyl isocyanate
2-isocyanato-1-methyl-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents