missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitrophenol, 97%

$49.01 - $49.01

Chemical Identifiers

CAS 5428-54-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00043909
InChI Key UMFDLIXUUJMPSI-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitrotoluene, 2-methy-5-nitrophenol, 2-methyl-5-nitro phenol, 2-methyl-5-nitro-phenol, 4-nitro-2-hydroxytoluene, 5-nitro-2-cresol, 5-nitro-2-methylphenol, 5-nitro-o-cresol, o-cresol, 5-nitro, phenol, 2-methyl-5-nitro
PubChem CID 93576
IUPAC Name 2-methyl-5-nitrophenol
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2047206
View Documents
Alfa Aesar™
B2047206
5g Each for $49.01
 
Specifications

Chemical Identifiers

5428-54-6
153.137
UMFDLIXUUJMPSI-UHFFFAOYSA-N
93576
CC1=C(C=C(C=C1)[N+](=O)[O-])O
C7H7NO3
MFCD00043909
2-hydroxy-4-nitrotoluene, 2-methy-5-nitrophenol, 2-methyl-5-nitro phenol, 2-methyl-5-nitro-phenol, 4-nitro-2-hydroxytoluene, 5-nitro-2-cresol, 5-nitro-2-methylphenol, 5-nitro-o-cresol, o-cresol, 5-nitro, phenol, 2-methyl-5-nitro
2-methyl-5-nitrophenol

Specifications

Phenol-like
116°C to 120°C
5428-54-6
MFCD00043909
2-hydroxy-4-nitrotoluene, 2-methy-5-nitrophenol, 2-methyl-5-nitro phenol, 2-methyl-5-nitro-phenol, 4-nitro-2-hydroxytoluene, 5-nitro-2-cresol, 5-nitro-2-methylphenol, 5-nitro-o-cresol, o-cresol, 5-nitro, phenol, 2-methyl-5-nitro
CC1=C(C=C(C=C1)[N+](=O)[O-])O
153.137
153.14
2-Methyl-5-nitrophenol
180°C (15mmHg)
5g
C7H7NO3
UN2446
UMFDLIXUUJMPSI-UHFFFAOYSA-N
2-methyl-5-nitrophenol
93576
97%
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID

EINECSNumber : 226-580-7

TSCA : Yes

Recommended Storage : Ambient temperatures