Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitroimidazole, 99%

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-nitroimidazole
CAS 88054-22-2
Formula Weight 127.1
MDL Number MFCD00151322
Beilstein 4032
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6061418

  SDS
alfa aesar™
H6061418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6061430

  SDS
alfa aesar™
H6061430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.1
Melting Point 251° to 255°C
Chemical Name or Material 2-Methyl-5-nitroimidazole
Physical Form Solid
Assay 99%
MDL Number MFCD00151322
CAS 88054-22-2
Beilstein 4032
Density 1.426g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 399°C
Quantity 50g

Applications
2-Methyl-4(5)-nitroimidazole was used to study the mechanism of both the alkaline and acidic hydrolysis of tinidazole. It is mainly used to synthesize intermediate of medicines such as Metronidazole and dimetridazole.

Solubility
Slightly soluble in ethanol, easily soluble in diluted acid and diluted alkali.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.