Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitrobenzothiazole, 98%

$125.81 - $534.91

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-nitrobenzothiazole
CAS 2941-66-4
Molecular Formula C8H6N2O2S
Formula Weight 194.21
MDL Number MFCD00602735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2535406

  SDS
alfa aesar™
B2535406
5g Each for $125.81

AAB2535414

  SDS
alfa aesar™
B2535414
25g Each for $534.91
Description & Specifications

Specifications

Formula Weight 194.21
Chemical Name or Material 2-Methyl-5-nitrobenzothiazole
TSCA TSCA
Assay 98%
Molecular Formula C8H6N2O2S
MDL Number MFCD00602735
CAS 2941-66-4
Health Hazard 1 Warning
Melting Point 132° to 134°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.