missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-nitrobenzonitrile, TCI America™

$55.00 - $165.00

Chemical Identifiers

CAS 939-83-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00017040
InChI Key XOSDYLFXPMFRGF-UHFFFAOYSA-N
Synonym benzonitrile, 2-methyl-5-nitro, 5-nitro-o-tolunitrile, 2-cyano-4-nitrotoluene, 2-methyl-5-nitrobenzenecarbonitrile, 3-cyano-4-methylnitrobenzene, 2-methyl-5-nitro-benzonitrile, 4-nitro-2-cyanotoluene, 5-nitro-ortho-tolunitrile, 5-nitro-2-methylbenzonitril, 6-methyl-3-nitrobenzonitrile
PubChem CID 350143
IUPAC Name 2-methyl-5-nitrobenzonitrile
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
M25705G missing translation for 'viewDocuments' TCI America
M25705G
5g Each for $55.00
 
M257025G missing translation for 'viewDocuments' TCI America
M257025G
25g Each for $165.00
 
Specifications

Chemical Identifiers

939-83-3
162.148
XOSDYLFXPMFRGF-UHFFFAOYSA-N
350143
CC1=C(C=C(C=C1)[N+](=O)[O-])C#N
C8H6N2O2
MFCD00017040
benzonitrile, 2-methyl-5-nitro, 5-nitro-o-tolunitrile, 2-cyano-4-nitrotoluene, 2-methyl-5-nitrobenzenecarbonitrile, 3-cyano-4-methylnitrobenzene, 2-methyl-5-nitro-benzonitrile, 4-nitro-2-cyanotoluene, 5-nitro-ortho-tolunitrile, 5-nitro-2-methylbenzonitril, 6-methyl-3-nitrobenzonitrile
2-methyl-5-nitrobenzonitrile

Specifications

175°C
106°C
939-83-3
MFCD00017040
XOSDYLFXPMFRGF-UHFFFAOYSA-N
2-methyl-5-nitrobenzonitrile
350143
Crystal-Powder at 20°C
2-Methyl-5-nitrobenzonitrile
Yellow
5g
C8H6N2O2
benzonitrile, 2-methyl-5-nitro, 5-nitro-o-tolunitrile, 2-cyano-4-nitrotoluene, 2-methyl-5-nitrobenzenecarbonitrile, 3-cyano-4-methylnitrobenzene, 2-methyl-5-nitro-benzonitrile, 4-nitro-2-cyanotoluene, 5-nitro-ortho-tolunitrile, 5-nitro-2-methylbenzonitril, 6-methyl-3-nitrobenzonitrile
CC1=C(C=C(C=C1)[N+](=O)[O-])C#N
162.148
162.15
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1780

TSCA : No