missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-5-nitrobenzoic acid, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 276570050
Specifications
2-Methyl-5-nitrobenzoic acid | |
Glass bottle | |
176°C to 180°C | |
1975-52-6 | |
100.0 | |
C8H7NO4 | |
MFCD00007371 | |
2-methyl-5-nitro benzoic acid, 2-methyl-5-nitro-benzoic acid, 2-methyl-5-nitrobenzoic acid, 5-nitro-2-methyl benzoic acid, 5-nitro-2-methylbenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, pubchem2244, rarechem al bo 0285 | |
DJRFJAVPROZZFL-UHFFFAOYSA-N | |
181.15 | |
181.15 | |
99+% |
Authentic | |
Yellow to Beige | |
5g | |
99.0 | |
99+% | |
CH3C6H3(NO2)CO2H | |
09, 471 | |
Solubility in water: insoluble | |
CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O | |
519683 | |
Crystalline Powder |
Chemical Identifiers
1975-52-6 | |
181.15 | |
DJRFJAVPROZZFL-UHFFFAOYSA-N | |
519683 |
C8H7NO4 | |
MFCD00007371 | |
2-methyl-5-nitro benzoic acid, 2-methyl-5-nitro-benzoic acid, 2-methyl-5-nitrobenzoic acid, 5-nitro-2-methyl benzoic acid, 5-nitro-2-methylbenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, pubchem2244, rarechem al bo 0285 | |
CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 217-829-