missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-nitrobenzoic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC276570010

 View more versions of this product

Catalog No. AC276570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methyl-5-nitrobenzoic acid
Glass bottle
176°C to 180°C
1975-52-6
100.0
C8H7NO4
MFCD00007371
2-methyl-5-nitro benzoic acid, 2-methyl-5-nitro-benzoic acid, 2-methyl-5-nitrobenzoic acid, 5-nitro-2-methyl benzoic acid, 5-nitro-2-methylbenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, pubchem2244, rarechem al bo 0285
DJRFJAVPROZZFL-UHFFFAOYSA-N
181.15
181.15
99+%
Authentic
Yellow to Beige
1g
99.0
99+%
CH3C6H3(NO2)CO2H
09, 471
Solubility in water: insoluble
CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O
519683
Crystalline Powder

Chemical Identifiers

1975-52-6
181.15
DJRFJAVPROZZFL-UHFFFAOYSA-N
519683
C8H7NO4
MFCD00007371
2-methyl-5-nitro benzoic acid, 2-methyl-5-nitro-benzoic acid, 2-methyl-5-nitrobenzoic acid, 5-nitro-2-methyl benzoic acid, 5-nitro-2-methylbenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, pubchem2244, rarechem al bo 0285
CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 217-829-

SDS Documents