missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitrobenzenesulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ A1476022

 View more versions of this product

Catalog No. AAA1476022



Specifications

Specifications

2-Methyl-5-nitrobenzenesulfonyl chloride
135°C to 137°C (0.7mmHg)
100g
C7H6ClNO4S
UN3261
2-methyl-5-nitrobenzene-1-sulfonyl chloride, 5-nitro-o-toluenesulfonyl chloride, 2-methyl-5-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methyl-5-nitro, 4-nitrotoluen-2-sulfochlorid, o-toluenesulfonyl chloride, 5-nitro, 4-nitrotoluen-2-sulfonylchlorid, 4-nitrotoluene-2-sulphonyl chloride, 4-nitrotoluen-2-sulfochlorid czech, 2-methyl-5-nitrophenylsulfonyl chloride
CC1=C(C=C(C=C1)[N+](=O)[O-])S(=O)(=O)Cl
235.638
235.65
>110°C (230°F)
42°C to 46°C
121-02-8
MFCD00051695
2697079
WPGVQDHXOUAJBW-UHFFFAOYSA-N
2-methyl-5-nitrobenzenesulfonyl chloride
8457
97%

Chemical Identifiers

121-02-8
235.638
WPGVQDHXOUAJBW-UHFFFAOYSA-N
8457
CC1=C(C=C(C=C1)[N+](=O)[O-])S(=O)(=O)Cl
C7H6ClNO4S
MFCD00051695
2-methyl-5-nitrobenzene-1-sulfonyl chloride, 5-nitro-o-toluenesulfonyl chloride, 2-methyl-5-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methyl-5-nitro, 4-nitrotoluen-2-sulfochlorid, o-toluenesulfonyl chloride, 5-nitro, 4-nitrotoluen-2-sulfonylchlorid, 4-nitrotoluene-2-sulphonyl chloride, 4-nitrotoluen-2-sulfochlorid czech, 2-methyl-5-nitrophenylsulfonyl chloride
2-methyl-5-nitrobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : XT8000000

Documents