missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitrobenzeneboronic acid, 96%

Manufacturer:  Alfa Aesar™ H53160MD

 View more versions of this product

Catalog No. AAH53160MD



Specifications

Specifications

2-Methyl-5-nitrobenzeneboronic acid
250mg
C7H8BNO4
2-methyl-5-nitrophenyl boronic acid, 2-methyl-5-nitrobenzeneboronic acid, 2-methyl-5-nitrophenyl boranediol, boronic acid,b-2-methyl-5-nitrophenyl, 5-nitro-2-methyl-phenylboronic acid, 2-dimethyl-3-nitrophenylboronic acid, 2-borono-4-nitrotoluene, acmc-2097th, 3-borono-4-methylnitrobenzene, 2-methyl-5-nitrophenyl-boronic acid
B(C1=C(C=CC(=C1)[N+](=O)[O-])C)(O)O
180.954
180.96
62°C to 66°C
100960-11-0
MFCD00757435
HRJFQIRIDFXMNX-UHFFFAOYSA-N
(2-methyl-5-nitrophenyl)boronic acid
2773513
96%

Chemical Identifiers

100960-11-0
180.954
HRJFQIRIDFXMNX-UHFFFAOYSA-N
2773513
B(C1=C(C=CC(=C1)[N+](=O)[O-])C)(O)O
C7H8BNO4
MFCD00757435
2-methyl-5-nitrophenyl boronic acid, 2-methyl-5-nitrobenzeneboronic acid, 2-methyl-5-nitrophenyl boranediol, boronic acid,b-2-methyl-5-nitrophenyl, 5-nitro-2-methyl-phenylboronic acid, 2-dimethyl-3-nitrophenylboronic acid, 2-borono-4-nitrotoluene, acmc-2097th, 3-borono-4-methylnitrobenzene, 2-methyl-5-nitrophenyl-boronic acid
(2-methyl-5-nitrophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents