Please login to your online account to display your discounted pricing

2-Methyl-5-nitroaniline, 96%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC364911000

 View more versions of this product

Catalog No. AC364911000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 152.15
Quantity 100g
Color Ochre to yellow-orange
Beilstein 12, 844
Melting Point 101° to 107°C
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula CH3C6H3(NO)nH
CAS 99-55-8
Synonym 2-Amino-4-nitrotoluene
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-5-nitroaniline
Infrared Spectrum Authentic
Name Note 96%
Assay Percent Range 96%
Molecular Formula C7H8N2O2
MDL Number MFCD00007741
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-Methyl-5-nitroaniline
Name Note 96%
CAS 99-55-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H8N2O2
Linear Formula CH3C6H3(NO)nH
Formula Weight 152.15
MDL Number MFCD00007741
Synonym 2-Amino-4-nitrotoluene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.

SDS & Safety Documents