missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-nitroaniline, 96%, ACROS Organics™

Chemical Identifiers

CAS 99-55-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007741
InChI Key DSBIJCMXAIKKKI-UHFFFAOYSA-N
Synonym 2-amino-4-nitrotoluene
PubChem CID 7444
ChEBI CHEBI:66891
IUPAC Name 2-methyl-5-nitroaniline
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC364911000 View Documents Thermo Fisher Scientific
AC364911000
100g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

99-55-8
152.15
DSBIJCMXAIKKKI-UHFFFAOYSA-N
7444
2-methyl-5-nitroaniline
C7H8N2O2
MFCD00007741
2-amino-4-nitrotoluene
CHEBI:66891
CC1=C(C=C(C=C1)[N+](=O)[O-])N

Specifications

Authentic
Ochre to Orange-Yellow
100g
95.0
95% min. (GC)
CH3C6H3(NO2)NH2
12, 844
Solubility in water: insoluble.
CC1=C(C=C(C=C1)[N+](=O)[O-])N
152.15
CHEBI:66891
Crystalline Powder
2-Methyl-5-nitroaniline, 96%
Glass bottle
104.0°C to 107.0°C
99-55-8
100.0
C7H8N2O2
MFCD00007741
2-amino-4-nitrotoluene
DSBIJCMXAIKKKI-UHFFFAOYSA-N
2-methyl-5-nitroaniline
7444
152.15
96%
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 202-765-8