Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-nitro-2H-indazole, 98+%

$153.69 - $641.88

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-nitro-2H-indazole
CAS 5228-48-8
Molecular Formula C8H7N3O2
Formula Weight 177.16
MDL Number MFCD00051809
Beilstein 167044
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2236003

  SDS
alfa aesar™
B2236003
1g Each for $153.69

AAB2236006

  SDS
alfa aesar™
B2236006
5g Each for $641.88
Description & Specifications

Specifications

Formula Weight 177.16
Assay ≥98%
CAS 5228-48-8
Molecular Formula C8H7N3O2
Melting Point 126° to 132°C
Chemical Name or Material 2-Methyl-5-nitro-2H-indazole
MDL Number MFCD00051809
Beilstein 167044
Quantity 1g