Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-5-(methylthio)furan, 99%

Chemical Identifiers

Chemical Name or Material 2-Methyl-5-(methylthio)furan
CAS 13678-59-6
Molecular Formula C7H14OS
Formula Weight 128.19
MDL Number MFCD01208018
UN Number UN1993
Synonym 2-Methyl-5-(methylmercapto)furan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2199804

  SDS
alfa aesar™
B2199804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2199809

  SDS
alfa aesar™
B2199809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.19
Chemical Name or Material 2-Methyl-5-(methylthio)furan
TSCA TSCA
Assay 99%
Molecular Formula C7H14OS
MDL Number MFCD01208018
CAS 13678-59-6
UN Number UN1993
Synonym 2-Methyl-5-(methylmercapto)furan
Health Hazard 1 Warning
Boiling Point 79° to 81°C (50mmHg)
Quantity 2g
Flash Point 50°C (122°F)
Odor Stench
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H226
Flammable liquid and vapour.