missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-5-isopropylaniline 93.0+%, TCI America™
$353.37 - $1060.10
Chemical Identifiers
CAS | 2051-53-8 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.24 |
MDL Number | MFCD00044503 |
InChI Key | YKWALWNGEXPARQ-UHFFFAOYSA-N |
Synonym | 2-Amino-p-cymene, 5-Isopropyl-2-methylaniline, 5-Isopropyl-o-toluidine |
PubChem CID | 137414 |
IUPAC Name | 2-methyl-5-(propan-2-yl)aniline |
SMILES | CC(C)C1=CC=C(C)C(N)=C1 |
Chemical Identifiers
2051-53-8 | |
149.24 | |
YKWALWNGEXPARQ-UHFFFAOYSA-N | |
137414 | |
CC(C)C1=CC=C(C)C(N)=C1 |
C10H15N | |
MFCD00044503 | |
2-Amino-p-cymene, 5-Isopropyl-2-methylaniline, 5-Isopropyl-o-toluidine | |
2-methyl-5-(propan-2-yl)aniline |
Specifications
2051-53-8 | |
C10H15N | |
1 g | |
YKWALWNGEXPARQ-UHFFFAOYSA-N | |
2-methyl-5-(propan-2-yl)aniline | |
137414 | |
≥93.0% (GC) | |
2-Methyl-5-isopropylaniline |
Yellow | |
MFCD00044503 | |
2-Amino-p-cymene, 5-Isopropyl-2-methylaniline, 5-Isopropyl-o-toluidine | |
CC(C)C1=CC=C(C)C(N)=C1 | |
149.24 | |
149.24 | |
Liquid |
Safety and Handling
TSCA : No