missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-4-trimethylsilyl-1-buten-3-yne, 97%

Manufacturer:  Alfa Aesar™ 4418904

 View more versions of this product

Catalog No. AA4418904



Specifications

Specifications

2-Methyl-4-trimethylsilyl-1-buten-3-yne
32°C (15mmHg)
18387-60-5
MFCD00190206
2-methyl-4-trimethylsilyl-1-buten-3-yne
CC(=C)C#C[Si](C)(C)C
138.285
138.28
97%
0.778 g/mL
2g
C8H14Si
UN1993
HRGBALJHGYAWBL-UHFFFAOYSA-N
trimethyl(3-methylbut-3-en-1-ynyl)silane
579646
Liquid

Chemical Identifiers

18387-60-5
138.285
HRGBALJHGYAWBL-UHFFFAOYSA-N
579646
CC(=C)C#C[Si](C)(C)C
C8H14Si
MFCD00190206
2-methyl-4-trimethylsilyl-1-buten-3-yne
trimethyl(3-methylbut-3-en-1-ynyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents