Please login to your online account to display your discounted pricing

2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-sulfonyl chloride, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC76303DA

 View more versions of this product

Catalog No. CC76303DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 265.66
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
Chemical Name or Material 2-Methyl-4-(trifluoromethyl)-1, 3-thiazole-5-sulfonyl chloride
Assay Percent Range 95%
Molecular Formula C5H3ClF3NO2S2
CAS 115151222
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-Methyl-4-(trifluoromethyl)-1, 3-thiazole-5-sulfonyl chloride
Assay 97%
Assay Percent Range 95%
Molecular Formula C5H3ClF3NO2S2
Formula Weight 265.66
Accessories

GHS Signal Word: Danger

SDS & Safety Documents