Please login to your online account to display your discounted pricing

2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-sulfonyl chloride, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyl-4-(trifluoromethyl)-1, 3-thiazole-5-sulfonyl chloride
Assay 97%
Assay Percent Range 95%
Molecular Formula C5H3ClF3NO2S2
Formula Weight 265.66
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC76303CB

  SDS
fisher scientific company
CC76303CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC76303DA

  SDS
fisher scientific company
CC76303DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 265.66
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
Chemical Name or Material 2-Methyl-4-(trifluoromethyl)-1, 3-thiazole-5-sulfonyl chloride
Assay Percent Range 95%
Molecular Formula C5H3ClF3NO2S2
CAS 115151222
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger