missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS
117724-63-7
Molecular Formula
C6H4F3NO2S
Molecular Weight (g/mol)
211.16
MDL Number
MFCD00173295
InChI Key
REKJPVUFKQYMHW-UHFFFAOYSA-N
Synonym
2-methyl-4-trifluoromethyl thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl thiazole-5-carboxylicacid, 2-methyl-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethylthiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-methyl-4-trifluoromethyl, 5-thiazolecarboxylicacid, 2-methyl-4-trifluoromethyl, acmc-209yjt, methyltrifluoromethylthiazolecarboxylicacid, pubchem11380
PubChem CID
1486080
IUPAC Name
2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
SMILES
CC1=NC(=C(S1)C(O)=O)C(F)(F)F
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
CAS
117724-63-7
Molecular Weight (g/mol)
211.16
InChI Key
REKJPVUFKQYMHW-UHFFFAOYSA-N
PubChem CID
1486080
SMILES
CC1=NC(=C(S1)C(O)=O)C(F)(F)F
Molecular Formula
C6H4F3NO2S
MDL Number
MFCD00173295
Synonym
2-methyl-4-trifluoromethyl thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl thiazole-5-carboxylicacid, 2-methyl-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethylthiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-methyl-4-trifluoromethyl, 5-thiazolecarboxylicacid, 2-methyl-4-trifluoromethyl, acmc-209yjt, methyltrifluoromethylthiazolecarboxylicacid, pubchem11380
IUPAC Name
2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
Specifications
CAS
117724-63-7
CAS Max %
100.0
Molecular Formula
C6H4F3NO2S
Packaging
Amber Glass Bottle
Quantity
250mg
SMILES
CC1=NC(=C(S1)C(O)=O)C(F)(F)F
Molecular Weight (g/mol)
211.16
Formula Weight
211.16
Chemical Name or Material
2-Methyl-4-(trifluoromethyl)-1, 3-thiazole-5-carboxylic acid
CAS Min %
96.5
Assay Percent Range
97%
MDL Number
MFCD00173295
Synonym
2-methyl-4-trifluoromethyl thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl thiazole-5-carboxylicacid, 2-methyl-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethyl-thiazole-5-carboxylic acid, 2-methyl-4-trifluoromethylthiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-methyl-4-trifluoromethyl, 5-thiazolecarboxylicacid, 2-methyl-4-trifluoromethyl, acmc-209yjt, methyltrifluoromethylthiazolecarboxylicacid, pubchem11380
InChI Key
REKJPVUFKQYMHW-UHFFFAOYSA-N
IUPAC Name
2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
PubChem CID
1486080
Percent Purity
97%
Safety and Handling
Safety and Handling
GHS H Statement: Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed. Causes skin irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
Product Certifications