Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-4-trifluoromethoxy-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-4-trifluoromethoxy-DL-phenylalanine
Formula Weight 263.21
MDL Number MFCD22201062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3433303

 
alfa aesar™
H3433303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH34333MD

 
alfa aesar™
H34333MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.21
Physical Form Solid
MDL Number MFCD22201062
Chemical Name or Material 2-Methyl-4-trifluoromethoxy-DL-phenylalanine
Assay 97%
Quantity 1g