Please login to your online account to display your discounted pricing

2-Methyl-4-piperazinoquinoline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyl-4-piperazinoquinoline
CAS 82241-22-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H17N3
Formula Weight 227.31
MDL Number MFCD01935745
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RH02136EA

 
fisher scientific company
RH02136EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RH02136CB

  SDS
fisher scientific company
RH02136CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.31
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C14H17N3
Assay Percent Range 97%
MDL Number MFCD01935745
Chemical Name or Material 2-Methyl-4-piperazinoquinoline
Health Hazard 1 GHS Signal Word: Danger
CAS 82241-22-3
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.