missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-4-phenylthiazole, TCI America™

$74.86 - $268.89

Chemical Identifiers

CAS 1826-16-0
Molecular Formula C10H9NS
Molecular Weight (g/mol) 175.249
MDL Number MFCD00489998
InChI Key XROORURTAQOYLW-UHFFFAOYSA-N
Synonym 2-methyl-4-phenylthiazole, 2-methyl-4-phenyl-thiazole, thiazole, 2-methyl-4-phenyl, thiazole,2-methyl-4-phenyl, #
PubChem CID 584377
IUPAC Name 2-methyl-4-phenyl-1,3-thiazole
SMILES CC1=NC(=CS1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28925G View Documents TCI America
M28925G
5g Each for $268.89
 
M28921G View Documents TCI America
M28921G
1g Each for $74.86
 
Specifications

Chemical Identifiers

1826-16-0
175.249
XROORURTAQOYLW-UHFFFAOYSA-N
584377
CC1=NC(=CS1)C2=CC=CC=C2
C10H9NS
MFCD00489998
2-methyl-4-phenylthiazole, 2-methyl-4-phenyl-thiazole, thiazole, 2-methyl-4-phenyl, thiazole,2-methyl-4-phenyl, #
2-methyl-4-phenyl-1,3-thiazole

Specifications

284°C
70°C
1826-16-0
MFCD00489998
XROORURTAQOYLW-UHFFFAOYSA-N
2-methyl-4-phenyl-1,3-thiazole
584377
Crystal-Powder at 20°C
2-Methyl-4-phenylthiazole
White-Yellow
5g
C10H9NS
2-methyl-4-phenylthiazole, 2-methyl-4-phenyl-thiazole, thiazole, 2-methyl-4-phenyl, thiazole,2-methyl-4-phenyl, #
CC1=NC(=CS1)C2=CC=CC=C2
175.249
175.25
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No