Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-4-phenylthiazole, 97%

$82.26 - $293.82

Chemical Identifiers

Chemical Name or Material 2-Methyl-4-phenylthiazole
CAS 1826-16-0
Molecular Formula C10H9NS
Formula Weight 175.25
MDL Number MFCD00489998
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5172803

  SDS
alfa aesar™
H5172803
1g Each for $82.26

AAH5172806

  SDS
alfa aesar™
H5172806
5g Each for $293.82
Description & Specifications

Specifications

Formula Weight 175.25
Melting Point 51° to 56°C
Quantity 1g
Molecular Formula C10H9NS
MDL Number MFCD00489998
Chemical Name or Material 2-Methyl-4-phenylthiazole
CAS 1826-16-0
Assay 97%