missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-4-phenyl-2-butanol, TCI America™

Manufacturer:  TCI America M0398500G

 View more versions of this product

Catalog No. M0398500G



Specifications

Specifications

2-Methyl-4-phenyl-2-butanol
500g
C11H16O
Dimethyl-beta-phenylethylcarbinol, 2-Phenylethyldimethylcarbinol
CC(C)(CCC1=CC=CC=C1)O
164.248
164.25
≥99.0% (GC)
White-Yellow
103-05-9
MFCD00004472
YXVSKJDFNJFXAJ-UHFFFAOYSA-N
2-methyl-4-phenylbutan-2-ol
7632
Crystal-Lump at 20°C

Chemical Identifiers

103-05-9
164.248
YXVSKJDFNJFXAJ-UHFFFAOYSA-N
7632
CC(C)(CCC1=CC=CC=C1)O
C11H16O
MFCD00004472
Dimethyl-beta-phenylethylcarbinol, 2-Phenylethyldimethylcarbinol
2-methyl-4-phenylbutan-2-ol
Safety and Handling

Safety and Handling

RTECSNumber : EL5785000

TSCA : Yes

Documents