missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-4-phenyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC42201DA
Specifications
2-Methyl-4-phenyl-1, 3-thiazole-5-carboxylic acid | |
97% | |
C11H8NO2S | |
1g | |
APXSARIYDLVPAY-UHFFFAOYSA-M | |
2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid | |
7131132 | |
97% |
32002-72-5 | |
Amber Glass Bottle | |
MFCD07348569 | |
2-methyl-4-phenylthiazole-5-carboxylic acid, 2-methy-4-phenylthiazol-5-ylcarboxylic acid, 2-methyl-4-phenyl-thiazole-5-carboxylic acid, 2-methyl-4-phenylthiazol-5-ylcarboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl, 5-thiazolecarboxylic acid, 2-methyl-4-phenyl | |
CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1 | |
218.25 | |
219.26 |
Chemical Identifiers
32002-72-5 | |
218.25 | |
APXSARIYDLVPAY-UHFFFAOYSA-M | |
7131132 | |
CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1 |
C11H8NO2S | |
MFCD07348569 | |
2-methyl-4-phenylthiazole-5-carboxylic acid, 2-methy-4-phenylthiazol-5-ylcarboxylic acid, 2-methyl-4-phenyl-thiazole-5-carboxylic acid, 2-methyl-4-phenylthiazol-5-ylcarboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl, 5-thiazolecarboxylic acid, 2-methyl-4-phenyl | |
2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid |