Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-4-nitrobenzoic acid, 97%

$99.00 - $328.47

Chemical Identifiers

Chemical Name or Material 2-Methyl-4-nitrobenzoic acid
CAS 1975-51-5
Molecular Formula C8H7NO4
Formula Weight 181.15
MDL Number MFCD00210697
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3133203

  SDS
alfa aesar™
H3133203
1g Each for $99.00

AAH3133206

  SDS
alfa aesar™
H3133206
5g Each for $328.47
Description & Specifications

Specifications

Formula Weight 181.15
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 1975-51-5
MDL Number MFCD00210697
Chemical Name or Material 2-Methyl-4-nitrobenzoic acid
Melting Point 152° to 155°C
Molecular Formula C8H7NO4
Quantity 1g
Odor Odorless
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Assay 97%
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.